Learn More
Tosoh Bioscience LLC TSKGEL SUPERAW4000 6X150
Supplier:  Tosoh Bioscience LLC 0019317
Encompass
Column, Size Exclusion Chromatography (SEC), Aqueous and Organic; Tosoh TSKgel SuperAW4000; Media: Polymer; Particle: 6 micrometer; 6mm I.D. x 15cm Length; Stainless Steel
Catalog No. 50-851-550