Learn More
Tosoh Bioscience LLC TSKGEL SUPERAW5000 6X150
Supplier:  Tosoh Bioscience LLC 0019318
Encompass
Column, Size Exclusion Chromatography (SEC), Aqueous and Organic; Tosoh TSKgel SuperAW5000; Media: Polymer; Particle: 7 micrometer; 6mm I.D. x 15cm Length; Stainless Steel
Catalog No. 50-851-551