Learn More
Tosoh Bioscience LLC TSKGEL SUPERAWM-H 6X150
Supplier:  Tosoh Bioscience LLC 0019320
Encompass
Column, Size Exclusion Chromatography (SEC), Aqueous and Organic; Tosoh TSKgel SuperAWM-H; Media: Mixed bed polymer; Particle: 9 micrometer; 6mm I.D. x 15cm Length; Stainless Steel
Catalog No. 50-851-553