Learn More
Tosoh Bioscience LLC TSKGEL SUPERH-H GRD 4.6X35
Supplier:  Tosoh Bioscience LLC 0018003
Encompass
Column, Guard; For Tosoh TSKgel SuperH-H columns; Media: Polymer; Particle: 4 micrometer; 4.6mm I.D. x 3.5cm Length
Catalog No. 50-851-471