Learn More
Tosoh Bioscience LLC TSKGEL SUPERH-MH 6X150
Supplier:  Tosoh Bioscience LLC 0018001
Encompass
Column, Gel Permeation Chromatography; Tosoh TSKgel SuperHM-H; Media: Polymer; Particle: 3 micrometer and 5 micrometer; 6mm I.D. x 15cm Length;Stainless Steel
Catalog No. 50-851-469