Learn More
Tosoh Bioscience LLC TSKGEL SUPERH2000 6X150
Supplier: Tosoh Bioscience LLC 0017991
Column, Gel Permeation Chromatography; Tosoh TSKgel SuperH2000; Media: Polymer; Particle: 3 micrometer; 6mm I.D. x 15cm Length; Stainless Steel