Learn More
Tosoh Bioscience LLC TSKGEL SUPERH2500 6X150
Supplier:  Tosoh Bioscience LLC 0017992
Encompass
Column, Gel Permeation Chromatography; Tosoh TSKgel SuperH2500; Media: Polymer; Particle: 3 micrometer; 6mm I.D. x 15cm Length; Stainless Steel
Catalog No. 50-851-460