Learn More
Tosoh Bioscience LLC TSKGEL SUPERH6000 6X150
Supplier:  Tosoh Bioscience LLC 0017996
Encompass
Column, Gel Permeation Chromatography; Tosoh TSKgel SuperH6000; Media: Polymer; Particle: 3 micrometer; 6mm I.D. x 15cm Length; Stainless Steel
Catalog No. 50-851-464