Learn More
Tosoh Bioscience LLC TSKGEL SUPERHZ-H GRD 4.6X20
Supplier:  Tosoh Bioscience LLC 0019668
Encompass
Column, Guard; For 4.6mm I.D. Tosoh TSKgel SuperHZM-H column; Media: Polymer; Particle: 10 micrometer; 4.6mm I.D. x 215cm Length
Catalog No. 50-851-576