Learn More
Tosoh Bioscience LLC TSKGEL SUPERHZM-H 4.6X150 10UM
Supplier:  Tosoh Bioscience LLC 0019664
Encompass
Column, Size Exclusion Chromatography (SEC), Aqueous and Organic; Tosoh TSKgel SuperHZM-H; Media: Polymer; Particle: 10 micrometer; 4.6mm I.D. x 15cm Length; Stainless Steel
Catalog No. 50-851-572