Learn More
Tosoh Bioscience LLC TSKGEL SUPERHZM-M 6X150
Supplier:  Tosoh Bioscience LLC 0019663
Encompass
Column, Size Exclusion Chromatography (SEC), Aqueous and Organic; Tosoh TSKgel SuperHZM-M; Media: Polymer; Particle: 3 and 5 micrometer; 6mm I.D. x 15cm Length; Stainless Steel
Catalog No. 50-851-571