Learn More
Tosoh Bioscience LLC TSKGEL SUPERHZM-N 6X150 3UM
Supplier:  Tosoh Bioscience LLC 0019661
Encompass
Column, Size Exclusion Chromatography (SEC), Aqueous and Organic; Tosoh TSKgel SuperHZM-N; Media: Polymer; Particle: 3 micrometer; 6mm I.D. x 15cm Length; Stainless Steel
Catalog No. 50-851-569