Learn More
Tosoh Bioscience LLC TSKGEL SUPERQ-5PW 21.5X150
Catalog No. 50851498 Shop All Tosoh Bioscience LLC Products
Encompass
Column, Ion exchange chromatography; Tosoh; TSKgel SuperQ-5PW; Matrix: polymethacrylate; Stainless-steel; Counter ion: Clsup- Particle size: 13u m; Pore size: 1000angstrom pH: 2.0 -12; 21.5mm I.D. x 15cm length
Catalog No. 50-851-498 Supplier Tosoh Bioscience LLC Supplier No. 0018387
Add to Cart
Add to Cart
Request a Quote
Column, Ion exchange chromatography; Tosoh; TSKgel SuperQ-5PW; Matrix: polymethacrylate; Stainless-steel; Counter ion: Clsup- Particle size: 13u m; Pore size: 1000angstrom pH: 2.0 -12; 21.5mm I.D. x 15cm length