Learn More
Tosoh Bioscience LLC TSKGEL SUPERQ-5PW 8X75
Supplier:  Tosoh Bioscience LLC 0018386
Encompass
Column, Ion exchange chromatography; Tosoh; TSKgel SuperQ-5PW; Matrix: polymethacrylate; Glass; Counter ion: Clsup- Particle size: 10u m; Pore size: 1000angstrom pH: 2.0 -12; 8mm I.D. x 7.5cm length
Catalog No. 50-851-497