Learn More
Tosoh Bioscience LLC SuperSW3000 HPLC Column
Supplier: Tosoh Bioscience LLC 0021845
Shipped in 0.05% NaN3 and 0.1 M Na2SO4 in 0.1 M phosphate buffer, pH 6.7. Phase diol, L × I.D. 30 cm × 1 mm, 4 um particle size.