Learn More
Tosoh Bioscience LLC TSKGEL SUPERSW3000 4.6X300
Supplier:  Tosoh Bioscience LLC 0018675
Encompass
Column, Gel Filtration Chromatography; Tosoh TSKgel SuperSW3000; Media: Silica; Particle: 4 micrometer; Pore Size: 250 angstrom; 4.6mm I.D. x 30cm Length; Stainless Steel
Catalog No. 50-851-512