Learn More
Tosoh Bioscience LLC TSKGEL TMS-250 4.6X75
Supplier:  Tosoh Bioscience LLC 0007190
Encompass
Column, Reversed Phase Chromatography; Tosoh TSKgel TMS-250; Media: Silica; Particle: 10 micrometer; Pore Size: 250 angstrom; 4.6mm I.D. x 7.5cm Length; Stainless Steel
Catalog No. 50-851-176