Learn More
Tosoh Bioscience LLC TSKGEL TOP-OFF PWXL G-DNA-PW
Supplier:  Tosoh Bioscience LLC 0008035
Encompass_Preferred
Media, Top-Off Resin; For Tosoh TSKgel PWxl and G-DNA-PW; Particle: 10 micrometer; Media: Polymer; 1gm
Catalog No. 50-851-256