Learn More
Tosoh Bioscience LLC TSKGEL UP-SW3000 4.6X300
Catalog No. 501049799 Shop All Tosoh Bioscience LLC Products
Encompass
LC Gel Permeation / Gel Filtration Columns, TSKgel UP-SW3000, 4.6mmID x 300mm, 2 um
Catalog No. 50-104-9799 Supplier Tosoh Bioscience LLC Supplier No. 0023448
LC Gel Permeation / Gel Filtration Columns, TSKgel UP-SW3000, 4.6mmID x 300mm, 2 um