Learn More
Tosoh Bioscience LLC TSKgel UP-SW Aggregate, 4.6 mm ID, 30 cm L, 3 um particle size
Catalog No. NC1849024 Shop All Tosoh Bioscience LLC Products
Encompass
TSKgel UP-SW Aggregate, the latest addition to the UP-SW series of silica-based size exclusion chromatography (SEC) columns, are specifically designed to facilitate the analysis of mAb aggregates, high molecular weight (MW) proteins and nucleic acids. These columns are 3 um, 30 nm pore size SEC analytical columns that have been designed with a higher exclusion limit than other TSKgel UP-SW columns.
Catalog No. NC1849024 Supplier Tosoh Bioscience LLC Supplier No. 0023524
TSKgel UP-SW Aggregate, the latest addition to the UP-SW series of silica-based size exclusion chromatography (SEC) columns, are specifically designed to facilitate the analysis of mAb aggregates, high molecular weight (MW) proteins and nucleic acids. These columns are 3 um, 30 nm pore size SEC analytical columns that have been designed with a higher exclusion limit than other TSKgel UP-SW columns.