Learn More
Tosoh Bioscience LLC TSKGEL UPSW3000 GUARDDC 4.6X20
Catalog No. 501049802 Shop All Tosoh Bioscience LLC Products
Encompass
LC Guard Column Direct Connect, TSKgel UP-SW3000, 4.6mmID x 20mm, 2 um (The guard column can be directly connected to the analytical column without tubing between the two columns.)
Catalog No. 50-104-9802 Supplier Tosoh Bioscience LLC Supplier No. 0023451
LC Guard Column Direct Connect, TSKgel UP-SW3000, 4.6mmID x 20mm, 2 um (The guard column can be directly connected to the analytical column without tubing between the two columns.)