Learn More
Tosoh Bioscience LLC TSKgel UP-SW Aggregate DC Guard Column for 4.6 mm ID column, 3 um, 2 cm L
Catalog No. NC1849027 Shop All Tosoh Bioscience LLC Products
Encompass
TSKgel UP-SW Aggregate DC Guard Column for 4.6 mm ID column can be directly connected to the TSKgel UP-SW Aggregate analytical column without tubing between the two columns. A male-type outlet endfitting on the guard column enables the direct connection to the screw-type endfitting of the analytical column.
Catalog No. NC1849027 Supplier Tosoh Bioscience LLC Supplier No. 0023527
TSKgel UP-SW Aggregate DC Guard Column for 4.6 mm ID column can be directly connected to the TSKgel UP-SW Aggregate analytical column without tubing between the two columns. A male-type outlet endfitting on the guard column enables the direct connection to the screw-type endfitting of the analytical column.