Learn More
Tosoh Bioscience LLC TSKGEL VMPAK-25 2X50 7UM
Catalog No. 50851615 Shop All Tosoh Bioscience LLC Products
Encompass
Column, Specialty; Application: Quarternary Amines; Tosoh TSKgel VMpak-25; Media: Polymer; Particle: 7 micrometer; 2mm I.D. x 5cm Length
Catalog No. 50-851-615 Supplier Tosoh Bioscience LLC Supplier No. 0020011
Column, Specialty; Application: Quarternary Amines; Tosoh TSKgel VMpak-25; Media: Polymer; Particle: 7 micrometer; 2mm I.D. x 5cm Length