Learn More
Sigma Aldrich Fine Chemicals Biosciences TSKgelR BioAssistR G6PW HP
Catalog No. 502905243
Encompass
A TSKgel G6000PW column in PEEK column hardware TSKgel BioAssist G6PW is for ultra-low sample adsorption during virus analysis. Use TSKgel G6000PW for the analysis of water-soluble polymers up to 8 000 000 Da molecular weight.
Catalog No. 50-290-5243 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 820024
A TSKgel G6000PW column in PEEK column hardware TSKgel BioAssist G6PW is for ultra-low sample adsorption during virus analysis. Use TSKgel G6000PW for the analysis of water-soluble polymers up to 8 000 000 Da molecular weight.