Learn More
Sigma Aldrich Fine Chemicals Biosciences TSKgelR SWXL Columns phase
Catalog No. 502908700
Encompass
BioAssist G2SWxl G3SWxl and G4Swxl are made of PEEK housing material to reduce sample absorption to stainless steel or glass. These columns have the same attributes as the corresponding stainless steel columns (TSKgel G2000SWxl G3000Swxl and G4000SWxl). Widely considered the premium columns for analyzing biopolymers by GFC TSKgel G2000SWxl columns feature high pore volume per unit column volume low sample adsorption and excellent column efficiency all contributing to unsurpassed sample resolution. With 5 micron particles and 125Ã… pores TSKgel G2000SWxl is an excellent choice for small proteins and peptide separations. The fractionation range is applicable for globular protein samples up to 150000 Da.
Catalog No. 50-290-8700 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 820027
BioAssist G2SWxl G3SWxl and G4Swxl are made of PEEK housing material to reduce sample absorption to stainless steel or glass. These columns have the same attributes as the corresponding stainless steel columns (TSKgel G2000SWxl G3000Swxl and G4000SWxl). Widely considered the premium columns for analyzing biopolymers by GFC TSKgel G2000SWxl columns feature high pore volume per unit column volume low sample adsorption and excellent column efficiency all contributing to unsurpassed sample resolution. With 5 micron particles and 125Ã… pores TSKgel G2000SWxl is an excellent choice for small proteins and peptide separations. The fractionation range is applicable for globular protein samples up to 150000 Da.