Learn More
Aviva Systems Biology Corp TSKS Antibody - N-terminal region (ARP53748_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53748_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TSKS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TSKS. Alternate names: TSKS1, TSSKS, STK22S1ApplicationIHC, WB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61113