Learn More
Aviva Systems Biology Corp TSLP Antibody - C-terminal region (ARP60913_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60913_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TSLP. It was validated on Western Blot. . Alternate names: ApplicationWB. Species reactivity: Dog, Horse, Human
Catalog No. 50-287-201