Learn More
ACROBiosystems ACROBiosystems Mouse TSLP Protein, His Tag
Catalog No. 501956556
Encompass_Preferred
Recombinant Protein;1MG;Mouse TSLP, His Tag (TSP-M52H8) is expressed from human 293 cells (HEK293). It contains AA Tyr 20 - Glu 140 (Accession # Q9JIE6-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.9 kDa. The protein migrates as 23-27 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6556 Supplier ACROBiosystems Supplier No. TSPM52H81MG
Recombinant Protein;1MG;Mouse TSLP, His Tag (TSP-M52H8) is expressed from human 293 cells (HEK293). It contains AA Tyr 20 - Glu 140 (Accession # Q9JIE6-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.9 kDa. The protein migrates as 23-27 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.