Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TSLP Human HEK293 100ug
SDP

Catalog No. 5000184197
Encompass_Preferred

Recombinant human thymic stromal lymphopoietin (TSLP) produced in HEK293 cells, purified to >95% and supplied as a lyophilized powder with a C-terminal 10xHis tag. The protein shows functional activity in binding assays (ED50 ≤5.6 ng/mL), has low endotoxin (<1 EU/μg), and is formulated for stability and long-term storage at -20°C; follow manufacturer reconstitution and storage recommendations.

  • Expressed in HEK293 cells with a C-terminal His tag.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Functional in binding assays with ED50 ≤5.6 ng/mL.
  • Low endotoxin level (<1 EU/μg).
  • Lyophilized formulation for stability and shipping.
  • Recommended storage at -20°C; aliquot to avoid freeze-thaw.

Catalog No. 50-001-84197 Supplier Medchemexpress LLC Supplier No. HYP70818100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant human thymic stromal lymphopoietin (TSLP) produced in HEK293 cells, purified to >95% and supplied as a lyophilized powder with a C-terminal 10xHis tag. The protein shows functional activity in binding assays (ED50 ≤5.6 ng/mL), has low endotoxin (<1 EU/μg), and is formulated for stability and long-term storage at -20°C; follow manufacturer reconstitution and storage recommendations.

  • Expressed in HEK293 cells with a C-terminal His tag.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Functional in binding assays with ED50 ≤5.6 ng/mL.
  • Low endotoxin level (<1 EU/μg).
  • Lyophilized formulation for stability and shipping.
  • Recommended storage at -20°C; aliquot to avoid freeze-thaw.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.