Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thymic stromal lymphopoietin | >90.0% | 100UG
SDP

Catalog No. 5000175714
Encompass_Preferred

Recombinant human thymic stromal lymphopoietin (TSLP) expressed in HEK293 cells with N-Myc and 6xHis tags. This cytokine stimulates T-cell-attracting chemokine release from monocytes, enhances CD11c+ dendritic cell maturation, and can activate mast cells. Supplied for research use only.

  • Human expression system provides native-like post-translational modifications.
  • N-Myc and 6xHis tags facilitate detection and purification.
  • Purity >90% by reducing SDS-PAGE ensures consistent performance.
  • Predicted molecular mass ~18.9 kDa; shows 20-30 kDa bands on SDS-PAGE.
  • Suitable for in vitro immunology and signaling assays.
  • Use in receptor binding, cell-based functional assays, and mast cell activation studies.
  • Quality and batch data available via certificate of analysis.

Catalog No. 50-001-75714 Supplier Medchemexpress LLC Supplier No. HYP700601100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human thymic stromal lymphopoietin (TSLP) expressed in HEK293 cells with N-Myc and 6xHis tags. This cytokine stimulates T-cell-attracting chemokine release from monocytes, enhances CD11c+ dendritic cell maturation, and can activate mast cells. Supplied for research use only.

  • Human expression system provides native-like post-translational modifications.
  • N-Myc and 6xHis tags facilitate detection and purification.
  • Purity >90% by reducing SDS-PAGE ensures consistent performance.
  • Predicted molecular mass ~18.9 kDa; shows 20-30 kDa bands on SDS-PAGE.
  • Suitable for in vitro immunology and signaling assays.
  • Use in receptor binding, cell-based functional assays, and mast cell activation studies.
  • Quality and batch data available via certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.