Learn More
Medchemexpress LLC TSLP Human HEK293 2ug
SDP
Catalog No. 5000184577
Encompass_Preferred
Recombinant human thymic stromal lymphopoietin (TSLP) produced in HEK293 cells with a C-terminal 10xHis tag. Supplied as a lyophilized powder, this cytokine is suitable for binding assays, functional ELISAs, and cell-based studies of TSLP-TSLPR signaling, and is provided with high purity and low endotoxin to support sensitive bioassays and reproducible results.
  • High purity greater than 95% as determined by reducing SDS-PAGE.
  • Glycosylated protein expressed in mammalian cells for native folding.
  • Includes defined residues Y29-Q159 corresponding to the mature isoform.
  • Lyophilized formulation for convenient storage and shipment.
  • Low endotoxin (<1 EU/μg) suitable for cell-based assays.
  • Demonstrated bioactivity with ED50 values in the ng/mL range.
  • Stable when stored at -20°C and after reconstitution with an appropriate carrier protein.
Catalog No. 50-001-84577 Supplier Medchemexpress LLC Supplier No. HYP708182UG
Recombinant human thymic stromal lymphopoietin (TSLP) produced in HEK293 cells with a C-terminal 10xHis tag. Supplied as a lyophilized powder, this cytokine is suitable for binding assays, functional ELISAs, and cell-based studies of TSLP-TSLPR signaling, and is provided with high purity and low endotoxin to support sensitive bioassays and reproducible results.
  • High purity greater than 95% as determined by reducing SDS-PAGE.
  • Glycosylated protein expressed in mammalian cells for native folding.
  • Includes defined residues Y29-Q159 corresponding to the mature isoform.
  • Lyophilized formulation for convenient storage and shipment.
  • Low endotoxin (<1 EU/μg) suitable for cell-based assays.
  • Demonstrated bioactivity with ED50 values in the ng/mL range.
  • Stable when stored at -20°C and after reconstitution with an appropriate carrier protein.