Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TSLP protein, human (HEK293, His-Myc) | >90.0% | 50UG
SDP

Catalog No. 5000177680
Encompass_Preferred

TSLP protein, human (HEK293, His-Myc) is a recombinant human thymic stromal lymphopoietin produced in HEK293 cells and provided lyophilized for in vitro research applications targeting TSLP signaling and immune cell activation.

  • Recombinant human cytokine with His-Myc tag.
  • Produced in HEK293 expression system.
  • Lyophilized formulation with trehalose for enhanced stability.
  • Available in small-scale sizes: 20 μg, 50 μg, 100 μg.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Predicted molecular mass ~18.9 kDa; observed SDS-PAGE bands ~20, 25, 30 kDa.

Catalog No. 50-001-75923 Supplier Medchemexpress LLC Supplier No. HYP70060150UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

TSLP protein, human (HEK293, His-Myc) is a recombinant human thymic stromal lymphopoietin produced in HEK293 cells and provided lyophilized for in vitro research applications targeting TSLP signaling and immune cell activation.

  • Recombinant human cytokine with His-Myc tag.
  • Produced in HEK293 expression system.
  • Lyophilized formulation with trehalose for enhanced stability.
  • Available in small-scale sizes: 20 μg, 50 μg, 100 μg.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Predicted molecular mass ~18.9 kDa; observed SDS-PAGE bands ~20, 25, 30 kDa.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.