Learn More
ACROBiosystems ACROBiosystems Human TSLP (R127A, R130A) Protein, Mouse IgG2a Fc Tag
Catalog No. 501956565
Encompass_Preferred
Recombinant Protein;1MG;Human TSLP (R127A, R130A), Mouse IgG2a Fc Tag (TSP-H5255) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession # Q969D9-1 (R127A, R130A)).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 41.6 kDa. The protein migrates as 50-58 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TSLP R, Fc Tag (Cat. No. TSR-H525a) at 5 μg/mL (100 μL/well) can bind Human TSLP (R127A, R130A), Mouse IgG2a Fc Tag (Cat. No. TSP-H5255) with a linear range of 0.3-1 ng/mL (QC tested).
Catalog No. 50-195-6565 Supplier ACROBiosystems Supplier No. TSPH52551MG
Recombinant Protein;1MG;Human TSLP (R127A, R130A), Mouse IgG2a Fc Tag (TSP-H5255) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession # Q969D9-1 (R127A, R130A)).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 41.6 kDa. The protein migrates as 50-58 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TSLP R, Fc Tag (Cat. No. TSR-H525a) at 5 μg/mL (100 μL/well) can bind Human TSLP (R127A, R130A), Mouse IgG2a Fc Tag (Cat. No. TSP-H5255) with a linear range of 0.3-1 ng/mL (QC tested).