Learn More
Medchemexpress LLC TSLP Rat HEK293 H 100ug | 100ug
SDP
Catalog No. 5000194014
Encompass_Preferred
TSLP protein as a cytokine plays a key role in immune regulation It induces monocytes to release T-cell attracting chemokines and significantly promotes the maturation of CD11c( ) dendritic cells TSLP Protein Rat (HEK293 His) is the recombinant rat-derived TSLP protein expressed by HEK293 with C-His labeled tag The total length of TSLP Protein Rat (HEK293 His) is 118 a a with molecular weight of 20-30 kDa
Catalog No. 50-001-94014 Supplier Medchemexpress LLC Supplier No. HYP78613100UG
TSLP protein as a cytokine plays a key role in immune regulation It induces monocytes to release T-cell attracting chemokines and significantly promotes the maturation of CD11c( ) dendritic cells TSLP Protein Rat (HEK293 His) is the recombinant rat-derived TSLP protein expressed by HEK293 with C-His labeled tag The total length of TSLP Protein Rat (HEK293 His) is 118 a a with molecular weight of 20-30 kDa