Learn More
ACROBiosystems Recombinant Protein;Human TSLP (R127A, R130A) Protein;HEK293;1MG;TSP,TSLP,TSLP R
Supplier:  ACROBiosystems TSPH52HA1MG
Encompass_Preferred
Recombinant Protein;Human TSLP (R127A, R130A) Protein;1MG;Human TSLP (R127A, R130A) (TSP-H52Ha) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession # Q969D9-1 (R127A, R130A)).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.7 kDa. The protein migrates as 22-27 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TSLP R, Fc Tag (Cat. No. TSR-H525a) at 5 μg/mL (100 μL/well) can bind Human TSLP (R127A, R130A) (Cat. No. TSP-H52Ha) with a linear range of 0.8-6 ng/mL (QC tested).;Immobilized Human TSLP (R127A, R130A) (Cat. No. TSP-H52Ha) at 1 μg/mL (100 μL/well) can bind Anti-TSLP, Human IgG2 with a linear range of 0.4-3 ng/mL (Routinely tested).
Catalog No. 50-165-7897