Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human TSLP, His Tag, Avi Tag (Avitag™);HEK293;25UG;TSP,TSLP,TSLP R
Supplier:  ACROBiosystems TSPH82EB25UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human TSLP, His Tag, Avi Tag (Avitag™);25UG;Biotinylated Human TSLP, His Tag, Avi Tag (TSP-H82Eb) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession # Q969D9-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 18.6 kDa. As a result of glycosylation, the protein migrates as 11 kDa and 18-21 kDa under reducing (R) condition, and 22-28 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human TSLP R, Fc Tag at 5 μg/mL (100 μL/well) can bind Biotinylated Human TSLP, His Tag, Avi Tag (Cat. No. TSP-H82Eb) with a linear range of 4-31 ng/mL (QC tested).;Immobilized Anti-TSLP, Human IgG2 at 2 μg/mL (100 μL/well) can bind Biotinylated Human TSLP, His Tag, Avi Tag (Cat. No. TSP-H82Eb) with a linear range of 0.1-2 ng/mL (Routinely tested).
Catalog No. 50-165-8055