Learn More
ACROBiosystems Recombinant Protein;Human TSLP R Protein, Fc Tag;HEK293;1MG;TSR,TSLP R,CRLF2,IL-XR,ILXR,p2RY8/CRLF2 fusion,TSLP receptor,TSLPR,CRL2,CRLF2Y,Cytokine receptor-like 2,TSLP,Thymic Stromal Lymphopoietin,
Catalog No. 501658042
Encompass_Preferred
Recombinant Protein;Human TSLP R Protein, Fc Tag;1MG;Human TSLP R, Fc Tag (TSR-H525a) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Lys 231 (Accession # Q9HC73-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50.4 kDa. The protein migrates as 62-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TSLP R, Fc Tag (Cat. No. TSR-H525a) at 5 μg/mL (100 μL/well) can bind Biotinylated Human TSLP, His Tag, Avi Tag (Cat. No. TSP-H82Eb) with a linear range of 4-31 ng/mL (QC tested).
Catalog No. 50-165-8042 Supplier ACROBiosystems Supplier No. TSRH525A1MG
Recombinant Protein;Human TSLP R Protein, Fc Tag;1MG;Human TSLP R, Fc Tag (TSR-H525a) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Lys 231 (Accession # Q9HC73-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50.4 kDa. The protein migrates as 62-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TSLP R, Fc Tag (Cat. No. TSR-H525a) at 5 μg/mL (100 μL/well) can bind Biotinylated Human TSLP, His Tag, Avi Tag (Cat. No. TSP-H82Eb) with a linear range of 4-31 ng/mL (QC tested).