Learn More
MilliporeSigma TSM PREP AGAR STRIPS 50/BX
Catalog No. NC9814836
Encompass
Modified Tryptic Soy Agar for determination of the total count of fastidious and sublethally damaged microorganisms. Contains neutralizers against disinfectants and growth supplements. For use with RCS Microbial Air Samplers. Store at 2-25 Deg.C. Packaged in EPS box, 50 units per box.
Catalog No. NC9814836 Supplier MilliporeSigma Supplier No. 1442400050
Modified Tryptic Soy Agar for determination of the total count of fastidious and sublethally damaged microorganisms. Contains neutralizers against disinfectants and growth supplements. For use with RCS Microbial Air Samplers. Store at 2-25 Deg.C. Packaged in EPS box, 50 units per box.