Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Asuragen Inc TSMIII Storage Buffer, 50 mL, 10 mM Tris, 100 mM NaCl, 1 mM MgCl2, 0.1% Gelatin, 0.3% Microcide III, pH 7.0, -15 to -30°C

Catalog No. NC1803665
Encompass

Asuragen TSMIII Storage Buffer is a specialized buffered solution designed to maintain stability for protein-coated nucleic acids. This reagent is ideal for the dilution of Asuragen's Armored products, ensuring the integrity of these sensitive biological materials through multiple freeze-thaw cycles. The stability provided by this storage buffer allows for flexible and scalable testing processes in various molecular assay applications.

  • Volume: 50 mL
  • Composition: 10 mM Tris, 100 mM NaCl, 1 mM MgCl2, 0.1% Gelatin, 0.3% Microcide III
  • pH: 7.0
  • Primary application: Dilution of Asuragen's Armored products
  • Key benefit: Provides stability to protein-coated nucleic acids
  • Storage stability: Withstands multiple freeze-thaw cycles
  • Recommended storage temperature: -15 to -30 °C (secondary)
  • Testing process benefit: Enables flexible and scalable testing processes

Catalog No. NC1803665 Supplier Asuragen Inc Supplier No. 49600
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Asuragen TSMIII Storage Buffer is a specialized buffered solution designed to maintain stability for protein-coated nucleic acids. This reagent is ideal for the dilution of Asuragen's Armored products, ensuring the integrity of these sensitive biological materials through multiple freeze-thaw cycles. The stability provided by this storage buffer allows for flexible and scalable testing processes in various molecular assay applications.

  • Volume: 50 mL
  • Composition: 10 mM Tris, 100 mM NaCl, 1 mM MgCl2, 0.1% Gelatin, 0.3% Microcide III
  • pH: 7.0
  • Primary application: Dilution of Asuragen's Armored products
  • Key benefit: Provides stability to protein-coated nucleic acids
  • Storage stability: Withstands multiple freeze-thaw cycles
  • Recommended storage temperature: -15 to -30 °C (secondary)
  • Testing process benefit: Enables flexible and scalable testing processes

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.