Learn More
Alkali Scientific TSN (Tryptic Sulfite Neomycin) Agar, 1 Kilogram
SDP
Supplier:  Alkali Scientific TSN002CP
Encompass_Preferred
TSN (Tryptic Sulfite Neomycin) Agar, 1 Kilogram, is designed for microbial detection, especially for isolating sulfur-reducing bacteria. This selective medium is widely used in laboratory applications to identify Clostridium species and other anaerobic bacteria. The agar is formulated for consistency and reproducibility, providing a reliable foundation for bacterial detection and differentiation in clinical and environmental settings.
Catalog No. 50-006-61816