Learn More
Electron Microscopy Sciences Tsp, 25"X15ft/Roll
SDP
Catalog No. 50949664
Encompass_Preferred
A thickness of 0.001" "PTFE" film bonded to a 0.008" self-adhesive vinyl gives your work surface good protection. Easy to use: cut to desired size, remove the backing, and press into place.
Catalog No. 50-949-664 Supplier Electron Microscopy Sciences Supplier No. 72612
A thickness of 0.001" "PTFE" film bonded to a 0.008" self-adhesive vinyl gives your work surface good protection. Easy to use: cut to desired size, remove the backing, and press into place.