Learn More
New England Biolabs, Inc. Tsp45I – 200 units
SDP
Catalog No. 50812288
Encompass
A restriction endonuclease that recognizes the sequence ^GTSAC_.
Catalog No. 50-812-288 Supplier New England Biolabs, Inc. Supplier No. R0583S
A restriction endonuclease that recognizes the sequence ^GTSAC_.