Learn More
Aviva Systems Biology Corp TSPAN12 Antibody - middle region (ARP46887_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46887_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TSPAN12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TSPAN12. Alternate names: NET-2, TM4SF12, EVR5, NET2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59429