Learn More
Aviva Systems Biology Corp TSPAN15 Antibody - C-terminal region (ARP46889_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46889_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TSPAN15. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TSPAN15. Alternate names: 2700063A19Rik, NET-7, TM4SF15, NET7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53497