Learn More
ABclonal Technology TSPAN15 Rabbit pAb
SDP
Supplier:  ABclonal Technology A143951000UL
Encompass_Preferred
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. The use of alternate polyadenylation sites has been found for this gene.
Catalog No. 50-001-08563