Learn More
Aviva Systems Biology Corp TSPAN17 Antibody - middle region (ARP42969_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42969_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TSPAN17. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TSPAN17. Alternate names: FBX23, FBXO23, MGC14859, MGC71255, TM4SF17ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-58378