Learn More
Aviva Systems Biology Corp TSPAN2 Antibody - middle region (ARP46643_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46643_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TSPAN2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TSPAN2. Alternate names: 6330415F13Rik, FLJ12082, RP4-666F24.2, TSN2, TSPAN-2, NET3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59370