Learn More
Aviva Systems Biology Corp TSPAN3 Antibody - middle region (ARP46642_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46642_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TSPAN3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TSPAN3. Alternate names: TM4-A, TM4SF8, TSPAN-3ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59369