Learn More
Aviva Systems Biology Corp TSPAN32 Antibody - middle region (ARP46636_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46636_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TSPAN32. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human TSPAN32. Alternate names: MGC22455, PHEMX, PHMX, TSSC6, ART1ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-59368