Learn More
Aviva Systems Biology Corp Tspan33 Antibody - C-terminal region (ARP55782_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55782_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Tspan33. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: RGD1560915ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-078